Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ GSK-3 Inhibitor IX
Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
10 mg
30 mg
Description
- Packaged Under Inert Gas
- Cell Permeable

Specifications
Specifications
Color | Purple |
Target Proteins | GSK-3α/β |
CAS | 667463-62-9 |
Molecular Formula | C16H10BrN3O2 |
Formula Weight | 356.2g/mol |
Solubility | DMSO |
Content And Storage | Protect from light |
Form | Solid |
Quantity | 1 mg |
Purity | 97% by HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction