Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Luciferase Inhibitor II

Catalog No. 1191145MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-911-45MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-911-45MG Supplier MilliporeSigma™ Supplier No. 1191145MG
Add to Cart
Add to Cart

The Luciferase Inhibitor II, also referenced under CAS 10205-56-8, controls the biological activity of Luciferase.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Green
Description 95% by HPLC
CAS 10205-56-8
Molecular Formula C15H14N2S
Formula Weight 254.4g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.