Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Luteolin

Catalog No. 4400255MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-002-55MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-002-55MG Supplier MilliporeSigma™ Supplier No. 4400255MG
Add to Cart
Add to Cart

An antioxidant flavonoid and a free radical scavenger.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 90% by HPLC
Target Proteins rat liver cytosolic glutathione S-transferase activity
CAS 491-70-3
Molecular Formula C15H10O6
Formula Weight 286.2g/mol
Solubility Aqueous Alkaline Solutions, DMSO, or EtOH
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.