Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MALT1 Inhibitor II, Mepazine Hydrochloride

Catalog No. 5005000001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-050-00001 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-050-00001 Supplier MilliporeSigma™ Supplier No. 5005000001
Add to Cart
Add to Cart

MALT1 Inhibitor II, Mepazine HCl, CAS 60-89-9, is a cell-permeable inhibitor of MALT1. Does not affect caspase-3 or -8 activities.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Grayish to Off-white
Formulation Supplied as a hydrochloride salt.
Description 97% by HPLC
Target Proteins MALT1
Molecular Formula C19H23ClN2S
Formula Weight 346.1g/mol
Solubility DMSO or H2O
Content And Storage Hygroscopic, Protect from light
Form Powder
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.