Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MAP Kinase Inhibitor Set I

Catalog No. 4441801SET Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-418-01SET 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-418-01SET Supplier MilliporeSigma™ Supplier No. 4441801SET
Add to Cart
Add to Cart

The MAP Kinase Inhibitor Set I controls the biological activity of MAP Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Contains 5 mg of the MEK inhibitor PD 98059 (Cat. No. 513000), 1 mg each of the MAP kinase inhibitors SB 202190 (Cat. No. 559388) and SB 203580 (Cat. No. 559389), and 1 mg of the negative control, SB 202474 (Cat. No. 559387). Supplied with a data sheet.
TRUSTED_SUSTAINABILITY

Specifications

Target Proteins p38 kinase
Content And Storage Protect from light
Form Solid
Quantity 1 Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.