Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MAS 7

Catalog No. 4448961MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-489-61MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-489-61MG Supplier MilliporeSigma™ Supplier No. 4448961MG
Add to Cart
Add to Cart

A cell-permeable, amphiphilic, mastoparan analog with five-fold greater potency than Mastoparan.

  • Peptide Sequence: H-Ile-Asn-Leu-Lys-Ala-Leu-Ala-Ala-Leu-Ala-Lys-Ala-Leu-Leu-NH2
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Description 98% by HPLC
Target Proteins α-subunit of G-proteins
CAS 145854-59-7
Molecular Formula C67H124N18O15
Formula Weight 1421.8g/mol
Solubility H2O
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.