Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MAT2A Inhibitor II, FIDAS-5

Catalog No. 5041730001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-417-30001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-417-30001 Supplier MilliporeSigma™ Supplier No. 5041730001
Add to Cart
Add to Cart

A cell-permeable fluorinated N,N-dialkylaminostilbene (FIDAS) analogue of FIDAS-3 (Cat

A cell-permeable fluorinated N,N-dialkylaminostilbene (FIDAS) analogue of FIDAS-3 (Cat. No. 681677) that competes against SAM for MAT2A binding and is >2-fold more potent than FIDAS-3 in inhibiting MAT2A-catalyzed SAM synthesis both in cell-free assays (IC50 = 2.1μM) and in LS174T colorectal cancer (CRC) cultures (3μM). FIDAS-5 at 3μM is shown to completely inhibit the proliferation of LS174T for up to 7 d in vitro and effectively suppress HT29 CRC tumor expansion in mice in vivo (20mg/kg/day p.o.).
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 98% by HPLC
Target Proteins MAT2A
Molecular Formula C15H13ClFN
Solubility DMSO
Content And Storage Protect from light
Form Semi-solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.