Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Mcl-1 Inhibitor II

Catalog No. 5080530001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-805-30001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-805-30001 Supplier MilliporeSigma™ Supplier No. 5080530001
Add to Cart
Add to Cart

A cell-permeable hydroxynaphthalenylthioacetate that targets Mcl-1 BH3-binding groove and selectively competes against 8nM FITC-AHA-BIM142-161 for Mcl-1, but not Bcl-xL, binding (IC50 = 310nM vs >40μM; [Mcl-1] & [Bcl-x

A cell-permeable hydroxynaphthalenylthioacetate that targets Mcl-1 BH3-binding groove and selectively competes against 8nM FITC-AHA-BIM142-161 for Mcl-1, but not Bcl-xL, binding (IC50 = 310nM vs >40μM; [Mcl-1] & [Bcl-xL] = 6.25nM), displaying little inhibitory potency toward CYP450 isoforms 1A2/2D6/3A4/2C9 (IC50 = >10μM). Antiproliferation potencies against 2 murine lymphoma cell lines (GI50 = 300 nM/ MCl1-1780 & 1.1μM/Bcl2-1863) and 11 human cancer lines of various origins (GI50 from 1.6 to 25μM) correlate well with intracellular BH3 domain peptide-binding capacity. Likewise, greater apoptosis induction is observed in MCl1-1780 than the highly Bcl-2/Bcl-xL primed Bcl2-1863 cultures upon drug treatment. Stability studies using murine and human liver microsome preparations reveal t 1/2 values of 40 min and 55 min, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
Target Proteins Mcl-1
Molecular Formula C23H24F3N3O
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.