Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Mcl-1 Inhibitor, MIM1

Catalog No. 44413025MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-413-025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-413-025MG Supplier MilliporeSigma™ Supplier No. 44413025MG
Add to Cart
Add to Cart

A cell-permeable thiazolyliminopyrogallol that selectively neutralizes Mcl-1, but not Bcl-XL , antiapoptotic activity by directly targeting Mcl-1 BH3-binding site, effectively competing against Bid BH3 sequence for Mcl-1 binding (IC50

A cell-permeable thiazolyliminopyrogallol that selectively neutralizes Mcl-1, but not Bcl-XL, antiapoptotic activity by directly targeting Mcl-1 BH3-binding site, effectively competing against Bid BH3 sequence for Mcl-1 binding (IC50 = 4.8μM). Shown to induce caspase 3/7 activation and cell death in Mcl-1-, but not Bcl-XL-, dependent B-ALL cultures (6% vs. 82% survival, respectively; 10μM). In comparison, ABT-737 (Cat. No. 197333) affects only Bcl-XL-, but not Mcl-1-, dependent B-ALL cells. Neither MIM1 nor ABT-737 significantly affects the survival of wild-type or Bax-/- Bak-/- MEF.
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 97% by HPLC
Target Proteins Mcl-1
Molecular Formula C17H21N3O3S
Formula Weight 347.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.