Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MDL-12,330A, Hydrochloride

Catalog No. 4442001MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-420-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-420-01MG Supplier MilliporeSigma™ Supplier No. 4442001MG
Add to Cart
Add to Cart

Cell-permeable and irreversible inhibitor of adenylate cyclase (IC50 = 250 μM).

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins Adenylate cyclase
CAS 40297-09-4
Molecular Formula C23H36N2·HCl
Formula Weight 377g/mol
Solubility DMSO, EtOH, or H2O
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.