Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Mecamylamine Hydrochloride

Catalog No. 5052070001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-520-70001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-520-70001 Supplier MilliporeSigma™ Supplier No. 5052070001
Add to Cart
Add to Cart

A potent, nonselective, and noncompetitive antagonist of nAChRs (IC50 = 640 nM

A potent, nonselective, and noncompetitive antagonist of nAChRs (IC50 = 640 nM, 2.5μM, 3.6μM and 6.9μM for α3β4, α4β2, α3β2 and α7 subtype nAChRs, respectively). Biologically viable admitted orally or systematically. Widely used in investigating the roles of central nAChRs in the brain functions (learning and memory, decision making, cognition) and disorders (addiction, Autisms, Tourette's syndrome, Schizophrenia and various cognitive and mood disorders).
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by TLC
Target Proteins nAChR
Molecular Formula C11H21N·HCl
Solubility H2O
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.