Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Membrane Traffic Inhibitor, A5

Catalog No. 44480510MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-480-510MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-480-510MG Supplier MilliporeSigma™ Supplier No. 44480510MG
Add to Cart
Add to Cart

The Membrane Traffic Inhibitor, A5 controls the biological activity of adaptor complex AP-1.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Yellow
Description 97% by HPLC
Molecular Formula C17H24N2O·HCl
Formula Weight 308.9g/mol
Solubility H2O
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.