Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Menin-MLL Interaction Inhibitor, MI-2-2

Catalog No. 44482510MG Shop All MilliporeSigma Products
Change view
Click to view available options
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
44-482-510MG 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-482-510MG Supplier MilliporeSigma™ Supplier No. 44482510MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A cell-permeable thienopyrimidine compound

A cell-permeable thienopyrimidine compound that competes against MLL (Mixed Lineage Leukemia) for menin binding (IC50 = 46 and 520 nM, respectively, against MBM1 sequence MLL4-15 and MLL4-43 containing both MBM1 & MBM2) by targeting menin MLL-binding pocket with high affinity ( K D = 22nM), effectively disrupting cellular menin and MLL-AF9 fusion interaction (by >90% at 12μM in MLL-AF9-expressing HEK293 cells). Shown to inhibit the oncogenic proliferation of four MLL translocation-harboring human leukemia cell lines (by >95% in 12 d at 6μM), but not non-MLL leukemia Kasumi-1 and HAL-01 lines (≤16% inhibition at 12μM), by downregulating MLL target genes expression via simultaneous hematopoietic differentiation induction and killing of non-differentiated population.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 99% by HPLC
Target Proteins Menin
Molecular Formula C17H20F3N5S2·HCl
Solubility DMSO
Content And Storage Hygroscopic, Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.