Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ α-(Methylamino)isobutyric Acid

Catalog No. 5061290001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-612-90001 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-612-90001 Supplier MilliporeSigma™ Supplier No. 5061290001
Add to Cart
Add to Cart

A glutamine transport inhibitor selective for Na+-dependent amino acid transport system A.

A glutamine transport inhibitor selective for Na+-dependent amino acid transport system A. Used in studying the functional roles of neurotransmissions mediated by amino acids.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by TLC
Target Proteins Glutamine transporter
Molecular Formula C5H11NO2
Solubility H2O
Content And Storage Protect from light
Form Solid
Quantity 250 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.