Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ α,β-methylene ATP

Catalog No. 5054190001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-541-90001 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-541-90001 Supplier MilliporeSigma™ Supplier No. 5054190001
Add to Cart
Add to Cart

A metabolically stable and selective P2X purinergic receptor agonist (pEC50

A metabolically stable and selective P2X purinergic receptor agonist (pEC50 = 5.81) that is shown to be more potent than ATP. Elicits contractions in ileum through the release of endogenous acetylcholine, which can be blocked by tetrodotoxin (∽1 mM). Shown to depolarize cells of the longitudinal muscle strips (∽ 200nM to 4μM) that reaches a peak within 2-3 min after application and then decays to a steady level.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins P2X
Molecular Formula C11H18N5O12P3·xLi+
Solubility H2O
Content And Storage Hygroscopic, Protect from light
Form Powder
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.