Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Methysergide Maleate

Catalog No. 5064190001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-641-90001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-641-90001 Supplier MilliporeSigma™ Supplier No. 5064190001
Add to Cart
Add to Cart

An antagonist of the 5-HT2B (pki

An antagonist of the 5-HT2B (pki = 8.0-9.4) and 5-HT2C (pki = 8.6-9.1) receptor. At the 5-HT1A and some other 5-HT receptors it serves as a partial agonist. It is known to have partial agonist effects on other 5-HT receptors as well. In neuronal and platelet culture, Methysergide blocks serotonin action with an IC50 = 18μM. Originally developed in the 1960′s as a migraine/cluster headache treatment, but discontinued due to significant physiological and pyschotropic side effects. Methysergide is still actively used in research as a 5-HT2 receptor antagonist in both in vivo and in vitro experiments.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 99% by HPLC
Target Proteins 5-HT
Molecular Formula C21H27N3O2·C4H4O4
Formula Weight 469.54g/mol
Solubility H2O
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.