Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Mevastatin, Sodium Salt

Catalog No. 4747055MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-470-55MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47-470-55MG Supplier MilliporeSigma™ Supplier No. 4747055MG
Add to Cart
Add to Cart

Carboxylate form of Mevastatin that is active in whole cells and in cell-free assays.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by TLC
Target Proteins HMG-CoA reductase
Molecular Formula C23H35O6·Na
Formula Weight 430.5g/mol
Solubility DMSO or EtOH
Content And Storage Hygroscopic
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.