Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Microcystin-LW, Microcystis aeruginosa

Catalog No. 47581825UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-581-825UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47-581-825UG Supplier MilliporeSigma™ Supplier No. 47581825UG
Add to Cart
Add to Cart

Analog of Microcystin-LR with Trp substituted in place of Arg. Believed to be more cell-permeable than other microcystins making it useful for biochemical studies in intact cells.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Description 95% by HPLC
Target Proteins Protein phosphatase 1 (PP1) and protein phosphatase 2A (PP2A)
Molecular Formula C54H72N8O12
Formula Weight 1025.2g/mol
Solubility DMSO or MeOH
Form Film
Quantity 25 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.