Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Midkine Inhibitor, iMDK

Catalog No. 5080520001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-805-20001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-805-20001 Supplier MilliporeSigma™ Supplier No. 5080520001
Add to Cart
Add to Cart

A cell-permeable imidazothiazolyl-chromenone compound that reduces cellular midkine/MDK protein level in H441 lung adenocarcinoma cells in a dose-dependent manner (by >90% in 48 h at 25nM) via a yet unidentified mechanism

A cell-permeable imidazothiazolyl-chromenone compound that reduces cellular midkine/MDK protein level in H441 lung adenocarcinoma cells in a dose-dependent manner (by >90% in 48 h at 25nM) via a yet unidentified mechanism. Shown to inhibit the viability of MDK-positive HEK293, H441, and H520 cells (by ≥60% at 500nM in 48 h), but not MDK-negative NHLF or HEK293 (by <15% at 500nM in 48 h), via apoptosis induction as a result of PI 3-K/AKT signaling inhibition. Intraperitoneal injection is reported to be efficacious in retarding H441 tumor expansion in mice (9mg/kg; 3X to 5X per wk) in vivo .
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by HPLC
Target Proteins Midkine
Molecular Formula C21H13FN2O2S
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.