Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ miR-372/3 Inhibitor, Neomycin-S

Catalog No. 5098870001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-988-70001 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-988-70001 Supplier MilliporeSigma™ Supplier No. 5098870001
Add to Cart
Add to Cart

A cell-permeable neomycin derived artificial nucleobase conjugate. Inhibits the production of oncogenic miRNAs 372 (IC50 = 2.4 μM) by binding to pre-miRNAs and interrupts processing by Dicer.

  • Packaged Under Inert Gas
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by HPLC
Target Proteins miRNA-272/273
Molecular Formula C39H60O14N12S
Solubility DMSO
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 2 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.