Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Mitochondrial Content Inducer, BRD6897

Catalog No. 5059840001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-598-40001 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-598-40001 Supplier MilliporeSigma™ Supplier No. 5059840001
Add to Cart
Add to Cart

A cell-permeable indolylphenylthienopyrimidinone compound that increases mitochondrial content without affecting the cell size (∽10μM). Reduces mitochondrial protein turnover without affecting any known transcriptional programs of mitochondrial biogenesis.

A cell-permeable indolylphenylthienopyrimidinone compound that increases mitochondrial content without affecting the cell size (∽10μM). Reduces mitochondrial protein turnover without affecting any known transcriptional programs of mitochondrial biogenesis. Reversibly increases uncoupled respiration in non-dividing cells (∽50% increase in 3T3-L1 and HUVEC at 10μM), but appears to be toxic to dividing cells. Shown to increase the expression of mitochondrial DNA encoded oxidative phosphorylation (OxPhos) genes without affecting nuclear DNA encoded OxPhos gene expression.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by HPLC
Molecular Formula C25H21N3O2S2
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.