Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Mitochondrial Pyruvate Carrier Inhibitor, UK5099

Catalog No. 5048170002 Shop All MilliporeSigma Products
Change view
Click to view available options
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-481-70002 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-481-70002 Supplier MilliporeSigma™ Supplier No. 5048170002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A cell-permeable α-cyanocinnamate compound that acts as a rapid, active site thiol modifying, time-dependent and non-competitive inhibitor of bi-directional pyruvate transport across plasma membranes in yeast, bacteria, Drosophila, and humans (IC50 = 50nM in rat heart mitochondria; Ki ≤ 1μM) with excellent selectivity over monocarboxylates and anions.

A cell-permeable α-cyanocinnamate compound that acts as a rapid, active site thiol modifying, time-dependent and non-competitive inhibitor of bi-directional pyruvate transport across plasma membranes in yeast, bacteria, Drosophila, and humans (IC50 = 50nM in rat heart mitochondria; Ki ≤ 1μM) with excellent selectivity over monocarboxylates and anions. Shown to significantly decrease pyruvate-driven respiration, enhance glucose uptake and activate AMPK (2μM in human myocytes and skeletal muscle myotubes).
TRUSTED_SUSTAINABILITY

Specifications

Color Bright Yellow
Description 99% by HPLC
Target Proteins mitochondria
Molecular Formula C18H12N2O2
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.