Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MLKL Inhibitor, Necrosulfonamide

Catalog No. 48007325MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-007-325MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-007-325MG Supplier MilliporeSigma™ Supplier No. 48007325MG
Add to Cart
Add to Cart

MLKL Inhibitor, Necrosulfonamide, CAS 432531-71-0, is a cell-permeable inhibitor that covalently modifies Cys88 and blocks human MLKL adaptor function.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Dark Yellow
Description 95% by HPLC
Target Proteins human MLKL
Molecular Formula C18H15N5O6S2
Formula Weight 461.5g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.