Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MMP-2 Inhibitor I

Catalog No. 44424410MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-424-410MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-424-410MG Supplier MilliporeSigma™ Supplier No. 44424410MG
Add to Cart
Add to Cart

The MMP-2 Inhibitor I, also referenced under CAS 10335-69-0, controls the biological activity of MMP-2. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by TLC
Target Proteins MMP-2
CAS 10335-69-0
Molecular Formula C18H35NO2
Formula Weight 297.5g/mol
Solubility EtOH or DMSO. Unstable in solution, reconstitute just prior to use
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.