Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MMP-2/MMP-9 Substrate I, Fluorogenic

Catalog No. 4442155MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-421-55MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-421-55MG Supplier MilliporeSigma™ Supplier No. 4442155MG
Add to Cart
Add to Cart

A fluorogenic substrate for both the 72 kDa human fibroblast gelatinase (HFG; MMP-2) and the 92 kDa human neutrophil gelatinase (HNG; MMP-9).

  • Packaged Under Inert Gas
  • Peptide Sequence: DNP-Pro-Leu-Gly-Met-Trp-Ser-Arg-OH
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Target Proteins A fluorogenic substrate for both human fibroblast gelatinase (HFG; MMP-2) and human neutrophil gelatinase (HNG; MMP-9).
Molecular Formula C44H61N13O13S
Formula Weight 1012.1g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.