Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MMP-2 Substrate, Fluorogenic

Catalog No. 4442121MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-421-21MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-421-21MG Supplier MilliporeSigma™ Supplier No. 4442121MG
Add to Cart
Add to Cart

A quenched fluorescence substrate for MMP-2 (gelatinase A; kcat/Km = 3.97 x 105 M-1s-1).

Peptide Sequence: MCA-Pro-Leu-Ala-Nva-Dpa-Ala-Arg-NH2 [Dpa = 3-(2,4-dinitrophenyl)-L-2,3-diaminopropionyl; Nva = L-norvaline]
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Target Proteins A quenched fluorescence substrate for MMP-2 (gelatinase A)
Molecular Formula C49H68N14O15
Formula Weight 1093.2g/mol
Solubility DMSO
Content And Storage Protect from Light, Protect from Moisture
Form Lyophilized
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.