Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MMP-3 Inhibitor I

Catalog No. 4442185MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-421-85MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-421-85MG Supplier MilliporeSigma™ Supplier No. 4442185MG
Add to Cart
Add to Cart

The MMP-3 Inhibitor I controls the biological activity of MMP-3. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Peptide Sequence: Ac-Arg-Cys-Gly-Val-Pro-Asp-NH2
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluroacetate salt.
Description 90% by HPLC
Target Proteins MMP-3
Molecular Formula C27H46N10O9S
Formula Weight 686.8g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.