Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MMP-3 Inhibitor II

Catalog No. 4442255MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-422-55MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-422-55MG Supplier MilliporeSigma™ Supplier No. 4442255MG
Add to Cart
Add to Cart

The MMP-3 Inhibitor II, also referenced under CAS 161314-17-6, controls the biological activity of MMP-3. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by TLC
Target Proteins human MMP-3
CAS 161314-17-6
Molecular Formula C13H20N2O5S
Formula Weight 316.4g/mol
Solubility DMSO or EtOH
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.