Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MMP Inhibitor III

Catalog No. 4442641MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-426-41MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-426-41MG Supplier MilliporeSigma™ Supplier No. 4442641MG
Add to Cart
Add to Cart

The MMP Inhibitor III, also referenced under CAS 927827-98-3, controls the biological activity of MMP. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 90% by HPLC
Target Proteins MMP-1
CAS 927827-98-3
Molecular Formula C19H29N3O4
Formula Weight 363.5g/mol
Solubility DMSO
Content And Storage Protect from moisture
Form Lyophilized
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.