Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MMP Substrate III, Fluorogenic

Catalog No. 444256500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-425-6500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-425-6500UG Supplier MilliporeSigma™ Supplier No. 444256500UG
Add to Cart
Add to Cart

A specific fluorogenic resonance energy transfer (FRET) peptide substrate with an MMP cleavable Gly-Leu bond and EDANS/Dabcyl as fluorophore/quencher combination.

  • Packaged Under Inert Gas
  • Peptide Sequence: DABCYL-GABA-Pro-Gln-Gly-Leu-Glu(EDANS)-Ala-Lys-NH2
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Formulation Supplied as a trifluoroacetate salt.
Description 97% by HPLC
Target Proteins Detection of MMP activity
Molecular Formula C63H88N16O14S
Formula Weight 1325.6g/mol
Solubility DMSO
Content And Storage Protect from light
Form Lyophilized
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.