Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Mouse Albumin

Catalog No. 12667425MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-667-425MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-667-425MG Supplier MilliporeSigma™ Supplier No. 12667425MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acts as stabilizing agent

  • Plasma protein
  • Lyophilized form
  • 30% Sterile-Filtered Aqueous Solution
  • Preservative free
  • Assay: ≥98% by SDS-PAGE
  • CAS: 9048-46-8

Recommended Storage

−20°C

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.