Learn More
Description
- Packaged Under Inert Gas
- Cell Permeable
- Deactivate with 5% Bleach
Specifications
Specifications
| Color | Light Beige |
| Description | 97% by HPLC |
| Product Type | Inhibitor |
| Molecular Formula | C10H8N2OS |
| Solubility | DMSO |
| Content And Storage | Protect from light |
| Form | Solid |
| Quantity | 25 mg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.