Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MPS1 Inhibitor III, AZ3146

Catalog No. 5319760001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-197-60001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-197-60001 Supplier MilliporeSigma™ Supplier No. 5319760001
Add to Cart
Add to Cart

A cell permeable purinone derivative that acts as a highly potent and selective inhibitor of monopolar spindle 1 kinase (MPS1; IC50 = ∽35

A cell permeable purinone derivative that acts as a highly potent and selective inhibitor of monopolar spindle 1 kinase (MPS1; IC50 = ∽35nM). Inhibits the phosphorylation of full-length immunoprecipitated human MPS1 and exhibits minimal activity against a panel of 49 other protein kinases. Has much reduced activity against FAK, JNK1, JNK2, and KIT and does not affect the activity of mitotic-specific phosphoforms of Aurora B and BubR1. Induces a significant reduction in time to complete mitosis (from 90 min to 32 min) and causes abnormal mitosis where over 50% of cells enter anaphase without aligning their chromosomes.

Please note that the molecular weight for this compound is batch-specific due to variable water content.
TRUSTED_SUSTAINABILITY

Specifications

Color Purple
Description 97% by HPLC
Target Proteins MPS1
Molecular Formula C24H32N6O3
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.