Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MT1-MMP Inhibitor, NSC405020

Catalog No. 44429525MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-429-525MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-429-525MG Supplier MilliporeSigma™ Supplier No. 44429525MG
Add to Cart
Add to Cart

The MT1-MMP Inhibitor, NSC405020 controls the biological activity of MT1-MMP. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by HPLC
Molecular Formula C12H15Cl2NO
Formula Weight 260.2g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.