Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MTEP Hydrochloride

Catalog No. 5046330001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-463-30001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-463-30001 Supplier MilliporeSigma™ Supplier No. 5046330001
Add to Cart
Add to Cart

A non-competitive, potent antagonist highly selective for mGlu5 receptors (IC50 =

A non-competitive, potent antagonist highly selective for mGlu5 receptors (IC50 = 5nM). Biologically active admitted orally or systematically. Widely used in assessing the functional roles of mGlu5 receptors in a variety of research areas, such as neuroprotection, depression, addiction, anxiety and stress related disorders, analgesia, locomotion, neural plasticity, and Parkinson's disease.
TRUSTED_SUSTAINABILITY

Specifications

Color Cream
Description 99% by HPLC
Target Proteins mGlu5 receptors
Molecular Formula C11H8N2SHCl
Solubility H2O
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.