Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ mTOR Activator, MHY1485

Catalog No. 5005540001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-055-40001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-055-40001 Supplier MilliporeSigma™ Supplier No. 5005540001
Add to Cart
Add to Cart

mTOR Activator, MHY1485, CAS Number: 326914-06-1, is a cell-permeable activator of mTOR that inhibits autophagy by suppressing the fusion between autophagosomes and lysosomes.

A cell-permeable triazine derivative that is shown to increase cellular mTOR Ser2448 and downstream substrate 4E-BP Thr37/46 phosphorylation level in rat liver Ac2F cells (1 to 2μM for 1 h), affecting culture viability only at much higher concentrations (by 20% after 24 h 20μM treatment). MHY1485 induced cellular LC3-II accumulation in Ac2F is reported to be a result of autophagy inhibition due to reduced fusion between autophagosomes and lysosomes. The mechanism of mTOR activation by MHY1485 is currently unknown.
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Yellow
Description 97% by HPLC
Molecular Formula C17H21N7O4
Formula Weight 387.39g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.