Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ mTOR Inhibitor XII, Torin2

Catalog No. 47599210MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-599-210MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47-599-210MG Supplier MilliporeSigma™ Supplier No. 47599210MG
Add to Cart
Add to Cart

A Torin1 (Cat

A Torin1 (Cat. No. 475991) structural analog that acts as a cell-permeable, potent, ATP-competitive inhibitor against DNA-PK and mTOR (IC50 = 0.5 and 2.81 nM, respectively). Comparing to Torin1, Torin2 displays enhanced inhibitory potency against PI 4-Kβ (IC50 = 18.3nM) and PI 3-K (IC50 =4.68, 5.67, 8.58, 17.5, 24.5, and 28.1 nM, respectively, against P110α/P85α, P110γ, hVPS34, P110δ/P85α, C2β, C2α, respectively), while being more effective in inhibiting S6K1 Thr389 phosphorylation in MEF cultures (IC50 = 0.25nM vs 2 nM, respectively, for Torin2 and Torin1) in vitro and displaying much improved oral bioavailability in mice ( C max (ng/mL) / T 1/2 (h) = 3968/0.72 and 223/0.79, respectively, for Torin2 and Torin1; 10 /mg/kg p.o.) in vivo .
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 99% by HPLC
Target Proteins MTOR
Molecular Formula C24H15F3N4O
Formula Weight 432.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.