Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Mucin, Bovine Submaxillary Gland

Catalog No. 499643500MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-964-3500MG 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 49-964-3500MG Supplier MilliporeSigma™ Supplier No. 499643500MG
Add to Cart
Add to Cart

Suitable as a substrate for neuraminidase. MilliporeSigma™; Calbiochem™ Mucin, Bovine Submaxillary Gland is a high molecular weight glycoprotein that is the major constituent of saliva, gastrointestinal juices and other secretions.

  • Mucin exists as an aggregated complex in excess of 106 molecular weight
  • Rich source of O-linked oligosaccharide chains
TRUSTED_SUSTAINABILITY

Specifications

Form Lyophilized
Quantity 500mg
Format Lyophilized, hygroscopic
Product Type Mucin, Bovine Submaxillary Gland
Storage Requirements Store at 2° to 8°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.