Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Myeloperoxidase Inhibitor-II

Catalog No. 5049080001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-490-80001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-490-80001 Supplier MilliporeSigma™ Supplier No. 5049080001
Add to Cart
Add to Cart

A 5-fluorotryptamine derivative that inhibits myeloperoxidase/MPO activity (IC50 = 18nM) via direct affinity interaction with enzyme active site heme pyrrole ring D, while exhibiting much reduced affinity toward serotonin reuptake transporter/SERT ( K i

A 5-fluorotryptamine derivative that inhibits myeloperoxidase/MPO activity (IC50 = 18nM) via direct affinity interaction with enzyme active site heme pyrrole ring D, while exhibiting much reduced affinity toward serotonin reuptake transporter/SERT ( K i = 631nM). Kinetic studies reveal that the inhibitor acts as a good one-electron donor for the intermediate Compound I Fe(IV)=O...Por+, facilitating the formation of Compound II (Fe(IV)-OH...Por) and thereby preventing the reduction of Compound I directly to the resting state Fe(IV)...Por via the halogenation cycle, while, on the other hand, behaves as a poor electron donor for Compound II, thereby preventing the completion of the peroxidase cycle by stalling the further reduction of Compound II to the resting state.

Please note that the molecular weight for this compound is batch-specific due to variable water content.
TRUSTED_SUSTAINABILITY

Specifications

Color Light Beige
Description 95% by HPLC
Target Proteins myeloperoxidase
Molecular Formula C12H13FN2O
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.