Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Myoseverin

Catalog No. 4759801MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-598-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47-598-01MG Supplier MilliporeSigma™ Supplier No. 4759801MG

missing translation for 'validMainframeMsg'

A microtubule-binding molecule that induces reversible fission of multinucleated myotubules into mononucleated fragments.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Description 95% by HPLC
Target Proteins Microtubule-binding molecule
CAS 267402-71-1
Molecular Formula C24H28N6O2
Formula Weight 432.5g/mol
Solubility DMSO
Content And Storage Protect from light
Form Viscous Gum
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.