Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Nα-Benzyloxycarbonyl-L-lysine Thiobenzyl Ester, Hydrochloride

Catalog No. 200274100MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-027-4100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-027-4100MG Supplier MilliporeSigma™ Supplier No. 200274100MG
Add to Cart
Add to Cart

Highly sensitive substrate for trypsin-like enzymes and granule-associated serine esterase.

Highly sensitive substrate for trypsin-like enzymes and granule-associated serine esterase.
TRUSTED_SUSTAINABILITY

Specifications

Color White to Off-white
Description >95% by HPLC
Target Proteins Substrate for trypsin-like enzymes and granule-associated serine esterase
Formula Weight 423.1g/mol
Solubility EtOH or H2O
Content And Storage Hygroscopic
Form Solid
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.