Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ N-Methyl-D-aspartic Acid, Hydrate

Catalog No. 454575100MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-457-5100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-457-5100MG Supplier MilliporeSigma™ Supplier No. 454575100MG
Add to Cart
Add to Cart

Excitatory amino acid neurotransmitter.

Excitatory amino acid neurotransmitter. Selective agonist of the glutamate receptor that regulates Ca2+ channels. Important in long-term potentiation, ischemia, and epilepsy. NMDA receptors are involved in the fine tuning of synaptic connections in the developing brain. Chronic treatment with NMDA produces structural changes in synaptic morphology. Over-excitation of NMDA receptors causes neuronal degeneration and cell death.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description >95% by TLC
Target Proteins Glutamate receptor that regulates Ca2+ channels
CAS 6384-92-5
Molecular Formula C5H9NO4·H2O
Formula Weight 165.1g/mol
Solubility H2O
Form Solid
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.