Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ NAAG

Catalog No. 48192050MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-192-050MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-192-050MG Supplier MilliporeSigma™ Supplier No. 48192050MG
Add to Cart
Add to Cart

NAAG is a neuropeptide and a neurotransmitter that is abundant in the mammalian nervous system.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by TLC
CAS 3106-85-2
Molecular Formula C11H16N2O8
Formula Weight 304.25g/mol
Solubility DMSO or MeOH
Content And Storage Protect from light
Form Powder
Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.