Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ NADP, Disodium Salt

Catalog No. 4819721GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-197-21GM 1 g
48-197-2100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 48-197-21GM Supplier MilliporeSigma™ Supplier No. 4819721GM
Add to Cart
Add to Cart

Participates in oxidation-reduction reactions

  • Coenzyme that is widely distributed in living matter
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Additive NADP, Disodium Salt
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.