Learn More
MilliporeSigma™ Calbiochem™ NADP, Monosodium Salt
Catalog No. 481971100MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-197-1100MG 100 mg
48-197-11GM 1 g
48-197-1500MG 500 mg
48-197-15GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 48-197-1100MG Supplier MilliporeSigma™ Supplier No. 481971100MG
Add to Cart
Add to Cart
Participates in oxidation-reduction reactions
  • Coenzyme that is widely distributed in living matter
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C
Additive NADP, Monosodium Salt
Quantity 100 mg