Learn More
MilliporeSigma™ Calbiochem™ α-Naphthyl Acid Phosphate, Monosodium Salt
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Color | White |
| Description | 98% by HPLC |
| Product Type | Phosphatase Inhibitor |
| Target Proteins | protein phosphatase |
| CAS | 81012-89-7 |
| Molecular Formula | C10H8O4PNa |
| Formula Weight | 246.1g/mol |
| Solubility | H2O |
| Content And Storage | Hygroscopic |
| Form | Solid |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.