Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ NBD-Binding Peptide II, Cell Permeable, CTP-NBD

Catalog No. 4800265MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-002-65MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-002-65MG Supplier MilliporeSigma™ Supplier No. 4800265MG
Add to Cart
Add to Cart

A cell-permeable cytoplasmic transduction peptide (CTP; YGRRARRRARR) fused to the NEMO binding domain peptide (NBD; TALDWSWLQTE) that acts as a potent inhibitor of TNFα-stimulated NF-κB activity (IC50∽ 50μM in HCT116 cells).

A cell-permeable cytoplasmic transduction peptide (CTP; YGRRARRRARR) fused to the NEMO binding domain peptide (NBD; TALDWSWLQTE) that acts as a potent inhibitor of TNFα-stimulated NF-κB activity (IC50 ∽ 50μM in HCT116 cells). Acts by interfering with the formation of active IKK complex and thereby prevents IκBα phosphorylation and its subsequent proteasomal degradation. Also blocks LPS-mediated increase in inducible nitric oxide synthase (iNOS) and cyclooxygenase-2 (COX-2) activities. Colon-targeted capsules containing CTP-NBD are shown to reduce 2,4,6-trinitrobenzenesulfonic acid (TNBS)-induced colitis in rodent models (4.5mg/kg/,p.o.) in vivo and diminish activities of myeloperoxidase and several inflammatory mediators. Antennapedia fused NBD cell-permeable peptide is also available ( Cat. No. 480025).
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Description 97% by HPLC
Target Proteins NEMO-binding peptide
Formula Weight 2846.2g/mol
Solubility H2O
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.