Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Necrostatin-1, Inactive Control

Catalog No. 4800665MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-006-65MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-006-65MG Supplier MilliporeSigma™ Supplier No. 4800665MG
Add to Cart
Add to Cart

A cell-permeable N-demethylated thiohydantoin analog of Nec-1 that is devoid of anti-necroptotic properties and serves as a suitable inactive control.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Light Yellow
Description 95% by HPLC
Target Proteins Inactive control for Nec-1 (Cat. No. 480065)
CAS 64419-92-7
Molecular Formula C12H11N3OS
Formula Weight 245.3g/mol
Solubility DMSO or MeOH
Content And Storage Protect from light
Form Crystalline Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.