Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Nefazodone Hydrochloride

Catalog No. 5081480001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-814-80001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-814-80001 Supplier MilliporeSigma™ Supplier No. 5081480001
Add to Cart
Add to Cart

A potent 5-HT2A receptor antagonist (Kd = 26nM), with moderate affinity for the alpha1-adrenergic receptor (Kd = 48nM) and 5-HT1A receptor

A potent 5-HT2A receptor antagonist (Kd = 26nM), with moderate affinity for the alpha1-adrenergic receptor (Kd = 48nM) and 5-HT1A receptor (Kd = 80nM). Originally developed as a clinical antidepressant but discontinued in 2004, nefazodone (Serzone) still remains a specific inhibitor of certain serotonergic pathways. Nefazodone also has been reported to induce mitochondrial dysfuntion.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by HPLC
Target Proteins 5-HT2A
Molecular Formula C25H32ClN5O2·HCl
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.