Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ NEMO-Binding Domain Binding Peptide, Cell-Permeable, Negative Control

Catalog No. 480030500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-003-0500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-003-0500UG Supplier MilliporeSigma™ Supplier No. 480030500UG
Add to Cart
Add to Cart

A cell-permeable Antennapedia-NBD mutated fusion peptide analog of NEMO-Binding Domain Binding Peptide that serves as a negative control.

  • Packaged Under Inert Gas
  • Peptide Sequence: H2N-Asp-Arg-Gln-Ile-Lys-Ile-Trp-Phe-Gln-Asn-Arg-Arg-Met-Lys-Trp-Lys-Lys-Thr-Ala-Leu-Asp-Ala-Ser-Ala-Leu-Gln-Thr-Glu-OH
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Target Proteins Negative control defective in binding to NEMO
Molecular Formula C154H249N47O42S
Formula Weight 3463g/mol
Solubility DMSO
Content And Storage Hygroscopic
Form Lyophilized
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.